siRNA Details
Gene Symbol: CDK11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313896.1 | NP_001300825.1 | 4 | 618 | ||
NM_001313982.1 | NP_001300911.1 | 4 | 618 | |||
NM_024011.3 | NP_076916.2 | 4 | 618 | |||
NM_033529.3 | NP_277071.2 | 4 | 618 | |||
NR_132739.1 | 4 | 618 | ||||
NR_132740.1 | 4 | 618 | ||||
GenBank mRNA | AF067518.1 | - | 4 | 376 | ||
AF067519.1 | - | 4 | 376 | |||
AF067520.1 | - | 4 | 376 | |||
AF067521.1 | - | 4 | 376 | |||
AF067522.1 | - | 4 | 376 | |||
AF067523.1 | - | 4 | 376 | |||
AF067524.1 | - | 1 | 45 | |||
AF067526.1 | - | 4 | 394 | |||
AF067527.1 | - | 4 | 394 | |||
AF067528.1 | - | 4 | 543 | |||
AF067529.1 | - | 4 | 543 | |||
AK301495.1 | - | 4 | 380 | |||
AK303446.1 | - | 4 | 376 | |||
AK303451.1 | - | 4 | 389 | |||
BC028187.1 | - | 4 | 497 | |||
DC388505.1 | - | 4 | 405 | |||
U07704.1 | - | 4 | 408 | |||
U07705.1 | - | 4 | 408 |
Gene Symbol: CDK11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF067512.1 | - | 4 | 408 | ||
AF067513.1 | - | 4 | 408 | |||
AF067514.1 | - | 5 | 455 | |||
AF067515.1 | - | 4 | 408 | |||
AF067516.1 | - | 5 | 455 | |||
AF067517.1 | - | 4 | 408 | |||
U04816.1 | - | 4 | 408 | |||
U04817.1 | - | 4 | 408 | |||
U04818.1 | - | 4 | 408 | |||
U04824.1 | - | 4 | 408 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
