siRNA Details
Gene Symbol: PI4KA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058004.3 | NP_477352.3 | 33 | 3943 | ||
XM_005261634.1 | XP_005261691.1 | 32 | 3850 | |||
XM_005261635.1 | XP_005261692.1 | 33 | 3943 | |||
XM_011530226.1 | XP_011528528.1 | 33 | 3943 | |||
XM_017028829.1 | XP_016884318.1 | 32 | 3877 | |||
XM_017028830.1 | XP_016884319.1 | 19 | 2099 | |||
GenBank mRNA | AB210002.1 | - | 33 | 3913 | ||
AF012872.1 | - | 32 | 3682 | |||
AK302424.1 | - | 33 | 3871 | |||
AK314781.1 | - | 5 | 597 | |||
BC018120.1 | - | 3 | 260 | |||
BC053654.1 | - | 2 | 236 | |||
L36151.1 | - | 1 | 229 |
Gene Symbol: PI4KAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003563.1 | 1 | 115 | |||
GenBank mRNA | AK096724.1 | - | 3 | 341 | ||
AK292695.1 | - | 2 | 206 | |||
BC072398.1 | - | 3 | 391 | |||
BC131538.1 | - | 1 | 115 |
Gene Symbol: PI4KAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003700.1 | 3 | 382 | |||
GenBank mRNA | AK126860.1 | - | 1 | 2346 | ||
AK298744.1 | - | 3 | 403 | |||
AK299734.1 | - | 3 | 247 | |||
BC020225.1 | - | 3 | 380 | |||
BC139841.1 | - | 1 | 103 | |||
BX452846.2 | - | 3 | 394 | |||
CR456378.1 | - | 1 | 121 | |||
DA463001.1 | - | 3 | 326 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
