siRNA Details
Gene Symbol: USP41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017029164.1 | XP_016884653.1 | 5 | 398 | ||
GenBank mRNA | AJ583822.1 | - | 4 | 371 | ||
AJ586979.1 | - | 3 | 308 | |||
AK303463.1 | - | 2 | 217 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
