siRNA Details
Gene Symbol: SDHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294332.1 | NP_001281261.1 | 11 | 1630 | ||
NM_004168.3 | NP_004159.2 | 12 | 1774 | |||
XM_011514072.1 | XP_011512374.1 | 12 | 1774 | |||
XM_017009685.1 | XP_016865174.1 | 12 | 1774 | |||
GenBank mRNA | AB208991.1 | - | 12 | 1660 | ||
AK094879.1 | - | 7 | 1595 | |||
AK131350.1 | - | 3 | 2388 | |||
AK131478.1 | - | 2 | 3864 | |||
AK225027.1 | - | 12 | 1675 | |||
AK291311.1 | - | 12 | 1673 | |||
AK295937.1 | - | 11 | 1535 | |||
AK302524.1 | - | 12 | 1681 | |||
BC001380.2 | - | 12 | 1670 | 3051442 | ||
BC041016.1 | - | 10 | 1278 | 3138724 | ||
D30648.1 | - | 12 | 1647 | |||
DQ402982.1 | - | 10 | 1446 | |||
L21936.1 | - | 12 | 1665 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
