siRNA Details
Gene Symbol: C2orf27A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013310.3 | NP_037442.3 | 4 | 897 | ||
GenBank mRNA | AF038169.1 | - | 4 | 897 | ||
AK309649.1 | - | 2 | 971 | |||
BC071972.1 | - | 4 | 565 | |||
BC093716.1 | - | 2 | 517 | 7939561 | ||
BC112189.1 | - | 2 | 517 | 8327657 |
Gene Symbol: C2orf27B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_214461.2 | NP_999626.1 | 3 | 1003 | ||
GenBank mRNA | BC043584.1 | - | 3 | 1003 | 5229449 |
Gene Symbol: LOC105372731 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017028519.1 | XP_016884008.1 | 2 | 714 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
