siRNA Details
Gene Symbol: PPP2R5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270476.1 | NP_001257405.1 | 4 | 549 | ||
NM_006245.3 | NP_006236.1 | 4 | 532 | |||
GenBank mRNA | AB000634.1 | - | 4 | 367 | ||
AB209861.1 | - | 2 | 111 | |||
AB451342.1 | - | 4 | 353 | |||
AB451357.1 | - | 4 | 353 | |||
AK122701.1 | - | 4 | 426 | |||
AK291797.1 | - | 4 | 440 | |||
AK299696.1 | - | 4 | 405 | |||
BC001095.2 | - | 4 | 416 | 3507280 | ||
BC010692.1 | - | 4 | 426 | 3853892 | ||
DA388594.1 | - | 4 | 532 | |||
L76702.1 | - | 4 | 540 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
