siRNA Details
Gene Symbol: ZNF214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013249.2 | NP_037381.2 | 3 | 2386 | ||
XM_005253128.3 | XP_005253185.1 | 4 | 2461 | |||
XM_005253129.3 | XP_005253186.1 | 3 | 2394 | |||
XM_006718308.3 | XP_006718371.1 | 3 | 2394 | |||
XM_006718309.3 | XP_006718372.1 | 3 | 2390 | |||
XM_011520355.2 | XP_011518657.1 | 4 | 2457 | |||
XM_011520357.2 | XP_011518659.1 | 3 | 2148 | |||
XM_017018269.1 | XP_016873758.1 | 3 | 2162 | |||
GenBank mRNA | AF056617.1 | - | 3 | 2341 | ||
BC136772.1 | - | 3 | 2266 | 9020762 | ||
BC144405.1 | - | 4 | 2325 | 9052926 |
