siRNA Details
Gene Symbol: RSC1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006511.2 | NP_006502.1 | 1 | 326 | ||
GenBank mRNA | AK314753.1 | - | 1 | 326 | ||
CR999543.1 | - | 1 | 101 |
Gene Symbol: DDI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032341.4 | NP_115717.3 | 10 | 1661 | ||
GenBank mRNA | AK057374.1 | - | 8 | 1057 | ||
AK293006.1 | - | 10 | 1661 | |||
AL598343.1 | - | 4 | 497 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
