siRNA Details
Gene Symbol: ASTN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014010.4 | NP_054729.3 | 6 | 1403 | ||
GenBank mRNA | AB014534.1 | - | 6 | 1403 | ||
AF116574.1 | - | 4 | 624 | |||
AK127350.1 | - | 5 | 636 | |||
AK315025.1 | - | 4 | 467 | |||
BC146756.1 | - | 6 | 1403 | 9007192 | ||
BX649176.1 | - | 6 | 738 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
