siRNA Details
Gene Symbol: GGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001560.2 | NP_001001560.1 | 5 | 725 | ||
NM_001172687.1 | NP_001166158.1 | 5 | 725 | |||
NM_001172688.1 | NP_001166159.1 | 6 | 771 | |||
NM_013365.4 | NP_037497.1 | 5 | 725 | |||
XM_005261517.3 | XP_005261574.1 | 6 | 950 | |||
XM_005261518.1 | XP_005261575.1 | 5 | 410 | |||
XM_005261520.1 | XP_005261577.1 | 5 | 616 | |||
XM_006724229.1 | XP_006724292.1 | 6 | 658 | |||
XM_011530122.1 | XP_011528424.1 | 6 | 883 | |||
XM_011530123.1 | XP_011528425.1 | 5 | 461 | |||
XM_011530124.1 | XP_011528426.1 | 6 | 950 | |||
XM_017028760.1 | XP_016884249.1 | 6 | 899 | |||
XM_017028761.1 | XP_016884250.1 | 4 | 2066 | |||
XM_017028762.1 | XP_016884251.1 | 6 | 899 | |||
XM_017028763.1 | XP_016884252.1 | 6 | 585 | |||
XM_017028764.1 | XP_016884253.1 | 5 | 616 | |||
GenBank mRNA | AF190862.1 | - | 5 | 353 | ||
AF218584.1 | - | 5 | 342 | |||
AF233521.1 | - | 5 | 360 | |||
AK075256.1 | - | 5 | 703 | |||
AK122898.1 | - | 6 | 771 | |||
AK225157.1 | - | 5 | 359 | |||
AK290548.1 | - | 5 | 364 | |||
AL110219.1 | - | 4 | 249 | |||
BC044629.1 | - | 5 | 354 | 5796877 | ||
CR456493.1 | - | 5 | 525 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
