siRNA Details
Gene Symbol: KCNIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013434.4 | NP_038462.1 | 2 | 243 | ||
GenBank mRNA | AB209076.1 | - | 2 | 229 | ||
AF120102.1 | - | 2 | 138 | |||
AF199599.1 | - | 1 | 128 | |||
AF367022.1 | - | 1 | 128 | |||
AJ131730.2 | - | 2 | 230 | |||
AK315437.1 | - | 2 | 189 | |||
BC012850.1 | - | 2 | 162 | 4179275 | ||
BI755669.1 | - | 2 | 275 | |||
BT020075.1 | - | 1 | 128 | |||
DQ148485.1 | - | 1 | 128 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
