siRNA Details
Gene Symbol: PILRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013439.2 | NP_038467.2 | 3, 4 | 893 | ||
GenBank mRNA | AF161080.1 | - | 3, 4 | 893 | ||
AJ400841.1 | - | 3, 4 | 834 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
