siRNA Details
Gene Symbol: DDR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202521.1 | NP_001189450.1 | 3 | 314 | ||
NM_001202522.1 | NP_001189451.1 | 3 | 314 | |||
NM_001202523.1 | NP_001189452.1 | 4 | 566 | |||
NM_001297652.1 | NP_001284581.1 | 4 | 685 | |||
NM_001297653.1 | NP_001284582.1 | 4 | 487 | |||
NM_001297654.1 | NP_001284583.1 | 4 | 487 | |||
NM_001954.4 | NP_001945.3 | 6 | 605 | |||
NM_013993.2 | NP_054699.2 | 5 | 531 | |||
NM_013994.2 | NP_054700.2 | 3 | 314 | |||
XM_006715185.2 | XP_006715248.1 | 4 | 478 | |||
XM_011514882.2 | XP_011513184.1 | 4 | 478 | |||
XM_011514883.1 | XP_011513185.1 | 6 | 593 | |||
XM_011514884.1 | XP_011513186.1 | 6 | 669 | |||
XM_011514885.1 | XP_011513187.1 | 4 | 516 | |||
XM_011514886.1 | XP_011513188.1 | 4 | 381 | |||
XM_011514887.1 | XP_011513189.1 | 4 | 531 | |||
XM_011514888.2 | XP_011513190.1 | 4 | 478 | |||
XM_017011268.1 | XP_016866757.1 | 4 | 348 | |||
XM_017011269.1 | XP_016866758.1 | 4 | 531 | |||
GenBank mRNA | AB210021.1 | - | 5 | 426 | ||
AK294793.1 | - | 4 | 552 | |||
BC008716.2 | - | 6 | 605 | 2964574 | ||
BC013400.2 | - | 6 | 605 | 2964574 | ||
BC070070.1 | - | 4 | 376 | 5262509 | ||
EU826613.1 | - | 3 | 272 | |||
EU826614.1 | - | 3 | 272 | |||
L20817.1 | - | 4 | 464 | |||
L57508.1 | - | 4 | 464 | |||
Z29093.1 | - | 3 | 608 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
