siRNA Details
Gene Symbol: GGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001560.2 | NP_001001560.1 | 8 | 1055 | ||
NM_001172687.1 | NP_001166158.1 | 8 | 1055 | |||
NM_001172688.1 | NP_001166159.1 | 9 | 1101 | |||
NM_013365.4 | NP_037497.1 | 8 | 1055 | |||
XM_005261517.3 | XP_005261574.1 | 9 | 1280 | |||
XM_005261518.1 | XP_005261575.1 | 8 | 740 | |||
XM_005261520.1 | XP_005261577.1 | 8 | 946 | |||
XM_005261522.1 | XP_005261579.1 | 3 | 209 | |||
XM_006724229.1 | XP_006724292.1 | 9 | 988 | |||
XM_011530122.1 | XP_011528424.1 | 9 | 1213 | |||
XM_011530123.1 | XP_011528425.1 | 8 | 791 | |||
XM_011530124.1 | XP_011528426.1 | 9 | 1280 | |||
XM_017028760.1 | XP_016884249.1 | 9 | 1229 | |||
XM_017028761.1 | XP_016884250.1 | 7 | 2396 | |||
XM_017028762.1 | XP_016884251.1 | 9 | 1229 | |||
XM_017028763.1 | XP_016884252.1 | 9 | 915 | |||
XM_017028764.1 | XP_016884253.1 | 8 | 946 | |||
XM_017028765.1 | XP_016884254.1 | 2 | 142 | |||
GenBank mRNA | AF190862.1 | - | 8 | 683 | ||
AF218584.1 | - | 8 | 672 | |||
AF233521.1 | - | 8 | 690 | |||
AK075256.1 | - | 8 | 1033 | |||
AK095427.1 | - | 3 | 206 | |||
AK122898.1 | - | 9 | 1101 | |||
AK225157.1 | - | 8 | 689 | |||
AK290548.1 | - | 8 | 694 | |||
AL110219.1 | - | 7 | 579 | |||
BC044629.1 | - | 8 | 684 | 5796877 | ||
CR456493.1 | - | 8 | 855 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
