siRNA Details
Gene Symbol: PPP2R3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013239.4 | NP_037371.2 | 8 | 1333 | ||
XM_011545630.2 | XP_011543932.1 | 8 | 1333 | |||
GenBank mRNA | AB209137.1 | - | 5 | 443 | ||
AF135016.1 | - | 7 | 697 | |||
AF155098.1 | - | 2 | 108 | |||
BC011180.1 | - | 2 | 110 | |||
BC063429.1 | - | 8 | 1088 | 5239974 | ||
BM715684.1 | - | 2 | 73 | |||
BU845905.1 | - | 11 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
