siRNA Details
Gene Symbol: ZBTB43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135776.1 | NP_001129248.1 | 2 | 3598 | ||
NM_014007.3 | NP_054726.1 | 3 | 3721 | |||
XM_005251835.3 | XP_005251892.1 | 3 | 4042 | |||
XM_011518409.1 | XP_011516711.1 | 4 | 3734 | |||
XM_011518411.2 | XP_011516713.1 | 2 | 3848 | |||
GenBank mRNA | AB007874.1 | - | 1 | 4579 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
