siRNA Details
Gene Symbol: TPTE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290224.1 | NP_001277153.1 | 5, 6 | 569 | ||
NM_199259.3 | NP_954868.1 | 9, 10 | 791 | |||
NM_199260.3 | NP_954869.1 | 8, 9 | 731 | |||
NM_199261.3 | NP_954870.2 | 10, 11 | 845 | |||
GenBank mRNA | AF007118.1 | - | 10, 11 | 793 | ||
AF495908.1 | - | 9, 10 | 739 | |||
AK308254.1 | - | 9, 10 | 687 | |||
AK314285.1 | - | 10, 11 | 744 | |||
AL833345.1 | - | 10, 11 | 752 | |||
AY219886.1 | - | 9, 10 | 739 | |||
AY219887.1 | - | 8, 9 | 679 | |||
AY219888.1 | - | 5, 6 | 517 | |||
BC028719.1 | - | 9, 10 | 774 | 5269598 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
