siRNA Details
Gene Symbol: TMEM176B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101311.1 | NP_001094781.1 | 6, 7 | 856 | ||
NM_001101312.1 | NP_001094782.1 | 5, 6 | 848 | |||
NM_001101314.1 | NP_001094784.1 | 4, 5 | 737 | |||
NM_014020.3 | NP_054739.3 | 5, 6 | 980 | |||
XM_006715933.3 | XP_006715996.2 | 5, 6 | 624 | |||
GenBank mRNA | AF115384.1 | - | 5, 6 | 770 | ||
AK000841.1 | - | 5, 6 | 679 | |||
AK097304.1 | - | 6, 7 | 1817 | |||
AK129525.1 | - | 4, 5 | 573 | |||
AK315575.1 | - | 6, 7 | 666 | |||
BC004358.1 | - | 5, 6 | 895 | 2822223 | ||
BC014586.2 | - | 5, 6 | 895 | 2822223 | ||
BC091471.1 | - | 5, 6 | 679 | |||
BI759037.1 | - | 4, 5 | 460 | |||
BI910761.1 | - | 6, 7 | 652 | |||
DQ144974.1 | - | 5, 6 | 738 |
