siRNA Details
Gene Symbol: OR11A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013937.3 | NP_039225.1 | 1 | 266 | ||
GenBank mRNA | AJ459857.1 | - | 1 | 266 | ||
BC131608.1 | - | 1 | 247 | 40114992 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
