siRNA Details
Gene Symbol: OSTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014028.3 | NP_054747.2 | 6 | 1213 | ||
GenBank mRNA | AF077205.1 | - | 4 | 566 | ||
AF533891.1 | - | 6 | 1197 | |||
AK075012.1 | - | 6 | 1139 | |||
AK129854.1 | - | 6 | 1207 | |||
AY358795.1 | - | 6 | 1204 | |||
BC068581.1 | - | 6 | 1195 | 5298450 |
