siRNA Details
Gene Symbol: OR10H3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013938.1 | NP_039226.1 | 1 | 803 | ||
GenBank mRNA | BC106707.2 | - | 1 | 803 | 40004023 | |
BC113844.1 | - | 1 | 803 | 40004027 |
Gene Symbol: OR10H4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004465.1 | NP_001004465.1 | 1 | 803 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
