siRNA Details
Gene Symbol: TF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF118093.1 | - | 1 | 1317 | ||
AK126941.1 | - | 1 | 2095 | |||
BX537660.1 | - | 1 | 437 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
