siRNA Details
Gene Symbol: CCDC82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017018307.1 | XP_016873796.1 | 1 | 225 | ||
GenBank mRNA | AF161533.1 | - | 1 | 263 | ||
AK313893.1 | - | 1 | 296 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
