siRNA Details
Gene Symbol: GTPBP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014170.2 | NP_054889.2 | 2 | 463 | ||
GenBank mRNA | AF161484.1 | - | 2 | 504 | ||
AK093419.1 | - | 2 | 1057 | |||
AK292651.1 | - | 2 | 482 | |||
BC037163.1 | - | 2 | 466 | 4620206 | ||
BG655089.1 | - | 2 | 397 | |||
BI830308.1 | - | 2 | 463 | |||
CK818022.1 | - | 2 | 384 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
