siRNA Details
Gene Symbol: YWHAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003404.4 | NP_003395.1 | 7 | 2210 | ||
NM_139323.3 | NP_647539.1 | 6 | 2115 | |||
XM_017028039.1 | XP_016883528.1 | 7 | 2014 | |||
GenBank mRNA | AF107406.1 | - | 1 | 975 | ||
AL834170.1 | - | 6 | 1994 | |||
BC010352.1 | - | 1 | 862 | |||
EU794580.1 | - | 7 | 2106 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
