siRNA Details
Gene Symbol: CPS1-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002763.2 | 1 | 2082 | |||
GenBank mRNA | AF090897.1 | - | 1 | 2091 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
