siRNA Details
Gene Symbol: CAMK2G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320898.1 | NP_001307827.1 | 20 | 2051 | ||
XM_011540225.2 | XP_011538527.1 | 19 | 1963 | |||
XM_017016716.1 | XP_016872205.1 | 21 | 2111 | |||
XM_017016717.1 | XP_016872206.1 | 21 | 2059 | |||
XM_017016718.1 | XP_016872207.1 | 20 | 2078 | |||
XM_017016719.1 | XP_016872208.1 | 21 | 2069 | |||
XM_017016720.1 | XP_016872209.1 | 20 | 2048 | |||
XM_017016721.1 | XP_016872210.1 | 21 | 2186 | |||
XM_017016722.1 | XP_016872211.1 | 20 | 2036 | |||
XM_017016723.1 | XP_016872212.1 | 19 | 2015 | |||
XM_017016724.1 | XP_016872213.1 | 19 | 2009 | |||
XM_017016725.1 | XP_016872214.1 | 19 | 1979 | |||
XM_017016726.1 | XP_016872215.1 | 19 | 1973 | |||
XM_017016727.1 | XP_016872216.1 | 18 | 1946 | |||
XM_017016729.1 | XP_016872218.1 | 19 | 1846 | |||
XM_017016731.1 | XP_016872220.1 | 19 | 1954 | |||
XM_017016732.1 | XP_016872221.1 | 19 | 1884 | |||
XM_017016734.1 | XP_016872223.1 | 19 | 1974 | |||
XM_017016737.1 | XP_016872226.1 | 18 | 1917 | |||
XM_017016738.1 | XP_016872227.1 | 20 | 1977 | |||
XM_017016739.1 | XP_016872228.1 | 21 | 2243 | |||
GenBank mRNA | AK131199.1 | - | 19 | 1947 |
Gene Symbol: DOT1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095280.1 | - | 4 | 3571 |
Gene Symbol: KCNQ1OT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002728.3 | 1 | 74317 |
Gene Symbol: KDM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097933.1 | - | 4 | 2796 |
Gene Symbol: LIMK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125156.1 | - | 2936 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
