siRNA Details
Gene Symbol: GTPBP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014170.2 | NP_054889.2 | 3, 4 | 618 | ||
NM_138485.1 | NP_612494.1 | 2, 3 | 519 | |||
GenBank mRNA | AF161484.1 | - | 3, 4 | 657 | ||
AK093419.1 | - | 3, 4 | 1212 | |||
AK289611.1 | - | 2, 3 | 481 | |||
AK292651.1 | - | 3, 4 | 637 | |||
BC037163.1 | - | 3, 4 | 621 | 4620206 | ||
BI830308.1 | - | 3, 4 | 618 | |||
BX354218.2 | - | 2, 3 | 487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
