siRNA Details
Gene Symbol: NDUFA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF044953.1 | - | 2 | 198 | ||
AK301520.1 | - | 2 | 241 | |||
BC001016.2 | - | 2 | 177 | 3345138 | ||
BU602971.1 | - | 2 | 275 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
