siRNA Details
Gene Symbol: GCAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014291.3 | NP_055106.1 | 1, 2 | 267 | ||
XM_005261411.1 | XP_005261468.1 | 1, 2 | 260 | |||
XM_017028676.1 | XP_016884165.1 | 1, 2 | 238 | |||
XM_017028677.1 | XP_016884166.1 | 1, 2 | 260 | |||
GenBank mRNA | AK290093.1 | - | 1, 2 | 260 | ||
BC014457.1 | - | 1, 2 | 224 | 4859130 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
