siRNA Details
Gene Symbol: FFAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005306.2 | NP_005297.1 | 1 | 963 | ||
XM_017026709.1 | XP_016882198.1 | 3 | 1502 | |||
XM_017026710.1 | XP_016882199.1 | 2 | 1379 | |||
XM_017026711.1 | XP_016882200.1 | 2 | 1137 | |||
GenBank mRNA | AB378083.1 | - | 1 | 963 | ||
AX810701.1 | - | 2 | 1043 | |||
BC095535.1 | - | 1 | 960 | |||
BC096198.3 | - | 1 | 963 | 40004248 | ||
BC096199.3 | - | 1 | 963 | 40004249 | ||
BC096200.1 | - | 1 | 963 | |||
BC096201.1 | - | 1 | 963 | 40004253 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:

Citations (1)
The imitation switch protein SNF2L regulates steroidogenic acute regulatory protein expression during terminal differentiation of ovarian granulosa cells.
- Authors
- Lazzaro MA, Pepin D, Pescador N, Murphy BD, Vanderhyden BC, Picketts DJ
- Journal
- Mol Endocrinol - Page No 2406 (Sun Oct 01 00:00:00 UTC 2006)
- Volume (Year)
- 20 (2006)
- Product Usage
- -
