siRNA Details
Gene Symbol: AMACR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167595.1 | NP_001161067.1 | 2, 3 | 495 | ||
NM_014324.5 | NP_055139.4 | 2, 3 | 495 | |||
GenBank mRNA | AF047020.1 | - | 2, 3 | 487 | ||
AF158378.1 | - | 2, 3 | 464 | |||
AJ130733.1 | - | 2, 3 | 487 | |||
AK000912.1 | - | 2, 3 | 480 | |||
AK022765.1 | - | 2, 3 | 465 | |||
AK222831.1 | - | 2, 3 | 467 | |||
AK225749.1 | - | 2, 3 | 504 | |||
AL545355.3 | - | 2, 3 | 408 | |||
AY330489.1 | - | 2, 3 | 408 | |||
AY935981.1 | - | 2, 3 | 408 | |||
BI550368.1 | - | 2, 3 | 533 | |||
DA156454.1 | - | 2, 3 | 498 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
