siRNA Details
Gene Symbol: THYN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037305.1 | NP_001032382.1 | 7, 8 | 740 | ||
NM_014174.2 | NP_054893.1 | 6, 7 | 1122 | |||
NM_199298.1 | NP_954995.1 | 7, 8 | 812 | |||
XM_005271527.3 | XP_005271584.1 | 7, 8 | 775 | |||
GenBank mRNA | AF059619.1 | - | 7, 8 | 784 | ||
AF087886.1 | - | 7, 8 | 745 | |||
AF161493.1 | - | 6, 7 | 1084 | |||
BC093074.1 | - | 7, 8 | 715 | 30717693 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
