siRNA Details
Gene Symbol: GCAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171690.1 | NP_001165161.1 | 7, 8 | 1135 | ||
NM_014291.3 | NP_055106.1 | 7, 8 | 1057 | |||
XM_005261410.3 | XP_005261467.1 | 7, 8 | 1136 | |||
XM_017028675.1 | XP_016884164.1 | 7, 8 | 1328 | |||
XM_017028677.1 | XP_016884166.1 | 7, 8 | 1050 | |||
GenBank mRNA | AF077740.1 | - | 7, 8 | 997 | ||
AK123190.1 | - | 7, 8 | 1103 | |||
AK290093.1 | - | 7, 8 | 1050 | |||
BC014457.1 | - | 7, 8 | 1014 | 4859130 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
