siRNA Details
Gene Symbol: CBX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303494.1 | NP_001290423.1 | 5 | 5153 | ||
NM_014292.4 | NP_055107.3 | 5 | 5207 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
