siRNA Details
Gene Symbol: HCAR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006018.2 | NP_006009.2 | 1 | 1267 | ||
GenBank mRNA | AK290930.1 | - | 1 | 1148 | ||
AK313212.1 | - | 1 | 1166 | |||
BC038955.1 | - | 1 | 1157 | |||
BC047891.1 | - | 1 | 1172 | 5750043 | ||
D10923.1 | - | 1 | 1172 |
Gene Symbol: HCAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177551.3 | NP_808219.1 | 1 | 1191 | ||
GenBank mRNA | AK290915.1 | - | 1 | 1130 | ||
AY148884.1 | - | 1 | 1177 | |||
BC027965.1 | - | 1 | 1177 | 5222688 | ||
BC056419.1 | - | 1 | 1163 | 5747743 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
