siRNA Details
Gene Symbol: ZNF324B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207395.2 | NP_997278.2 | 4 | 373 | ||
XM_005258915.4 | XP_005258972.1 | 5 | 446 | |||
XM_005258918.4 | XP_005258975.1 | 4 | 397 | |||
GenBank mRNA | AK127750.1 | - | 4 | 373 | ||
AK131452.1 | - | 3 | 1168 | |||
BC140881.1 | - | 4 | 366 | 9021749 | ||
BC140882.1 | - | 5 | 446 | 9021750 | ||
BX648711.1 | - | 4 | 346 |
Gene Symbol: ZNF324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014347.2 | NP_055162.1 | 4 | 411 | ||
XM_005258713.4 | XP_005258770.1 | 4 | 1206 | |||
GenBank mRNA | AF060503.1 | - | 4 | 360 | ||
AK092341.1 | - | 4 | 762 | |||
BC007717.2 | - | 4 | 364 | 4303415 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
