siRNA Details
Gene Symbol: CCT8L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014406.4 | NP_055221.1 | 1 | 461 | ||
GenBank mRNA | BC033797.1 | - | 1 | 461 | 5172617 | |
BC100811.2 | - | 1 | 327 | 40000853 | ||
BC100812.2 | - | 1 | 327 | 40000855 | ||
BC100813.1 | - | 1 | 327 | 40000856 | ||
BC139842.1 | - | 1 | 220 | |||
CR456508.1 | - | 1 | 316 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
