siRNA Details
Gene Symbol: KIR2DS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291695.1 | NP_001278624.1 | 6, 7 | 855 | ||
NM_001291696.1 | NP_001278625.1 | 5, 6 | 555 | |||
NM_001291700.1 | NP_001278629.1 | 4, 5 | 261 | |||
NM_001291701.1 | NP_001278630.1 | 5, 6 | 804 | |||
NM_012312.4 | NP_036444.1 | 6, 7 | 855 | |||
GenBank mRNA | AF074087.1 | - | 4, 5 | 217 | ||
AF285438.1 | - | 5, 6 | 780 | |||
AJ002102.1 | - | 6, 7 | 817 | |||
AJ002103.1 | - | 6, 7 | 820 | |||
AJ002104.1 | - | 5, 6 | 769 | |||
AJ002105.1 | - | 5, 6 | 769 | |||
AM409094.1 | - | 6, 7 | 821 | |||
AY366243.1 | - | 6, 7 | 830 | |||
AY366244.1 | - | 6, 7 | 819 | |||
BC108917.1 | - | 6, 7 | 822 | 40017299 | ||
BC119631.1 | - | 6, 7 | 839 | 40115568 | ||
BC119632.1 | - | 6, 7 | 839 | 40115570 | ||
BC125146.1 | - | 5, 6 | 804 | |||
BC126856.1 | - | 6, 7 | 822 | 40017301 | ||
L41347.1 | - | 6, 7 | 839 | |||
L76667.1 | - | 6, 7 | 537 | |||
L76668.1 | - | 5, 6 | 531 | |||
U24079.1 | - | 6, 7 | 818 | |||
X89893.1 | - | 6, 7 | 820 |
Gene Symbol: KIR3DL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322168.1 | NP_001309097.1 | 7, 8 | 1168 | ||
NM_013289.2 | NP_037421.2 | 7, 8 | 1168 | |||
GenBank mRNA | AF022049.1 | - | 7, 8 | 1105 | ||
AF103010.1 | - | 7, 8 | 1105 | |||
AF103011.1 | - | 7, 8 | 1105 | |||
AF103012.1 | - | 7, 8 | 1105 | |||
AF103013.1 | - | 7, 8 | 1105 | |||
AF262968.1 | - | 5, 6 | 1024 | |||
AF262969.1 | - | 5, 6 | 1024 | |||
AF262970.1 | - | 5, 6 | 1024 | |||
AF262971.1 | - | 5, 6 | 1024 | |||
AF262972.1 | - | 5, 6 | 1024 | |||
AF262973.1 | - | 7, 8 | 1138 | |||
AF262974.1 | - | 7, 8 | 1138 | |||
AJ417556.1 | - | 5, 6 | 1054 | |||
AJ417557.1 | - | 5, 6 | 1054 | |||
AY366236.1 | - | 7, 8 | 1143 | |||
AY366249.1 | - | 7, 8 | 1105 | |||
AY366250.1 | - | 7, 8 | 1131 | |||
AY366251.1 | - | 7, 8 | 1105 | |||
AY366252.1 | - | 7, 8 | 1105 | |||
AY760050.1 | - | 7, 8 | 1105 | |||
AY789065.1 | - | 7, 8 | 1120 | |||
AY789066.1 | - | 7, 8 | 1105 | |||
AY789072.1 | - | 7, 8 | 1105 | |||
BC028206.1 | - | 7, 8 | 1132 | 5215674 | ||
BC101540.1 | - | 7, 8 | 1124 | 8069046 | ||
BC101542.1 | - | 7, 8 | 1124 | 8069048 | ||
DQ178659.1 | - | 7, 8 | 1105 | |||
EF462885.1 | - | 5, 6 | 1035 | |||
EF472674.1 | - | 5, 6 | 1035 | |||
EF472675.1 | - | 5, 6 | 1035 | |||
EF472676.1 | - | 5, 6 | 1035 | |||
EF472677.1 | - | 5, 6 | 1035 | |||
EF472678.1 | - | 5, 6 | 1035 | |||
EF472679.1 | - | 5, 6 | 1035 | |||
L41269.1 | - | 7, 8 | 1124 | |||
L76664.1 | - | 6, 7 | 831 | |||
U30273.1 | - | 7, 8 | 1107 | |||
U30274.1 | - | 7, 8 | 1168 | |||
U31416.1 | - | 7, 8 | 1130 | |||
U33328.1 | - | 6, 7 | 1079 | |||
X94262.1 | - | 7, 8 | 1106 |
Gene Symbol: KIR3DS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | L76661.1 | - | 8, 9 | 1116 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
