siRNA Details
Gene Symbol: TPSB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024164.5 | NP_077078.5 | 1, 2 | 30 | ||
GenBank mRNA | AK308208.1 | - | 1, 2 | 30 | ||
BC029356.1 | - | 1, 2 | 30 | 4695458 | ||
S55551.1 | - | 1 | 14 |
Gene Symbol: TPSAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003294.3 | NP_003285.2 | 1, 2 | 59 | ||
GenBank mRNA | BC038416.1 | - | 1 | 13 | ||
BC074974.2 | - | 1, 2 | 40 | 30915512 | ||
M37488.1 | - | 1 | 12 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
