siRNA Details
Gene Symbol: TPSB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024164.5 | NP_077078.5 | 5 | 578 | ||
GenBank mRNA | AK308208.1 | - | 5 | 579 | ||
BC028059.1 | - | 4 | 567 | 5181815 | ||
BC029356.1 | - | 5 | 578 | 4695458 | ||
FJ931119.1 | - | 3 | 409 | |||
FJ931120.1 | - | 3 | 408 | |||
M33492.1 | - | 4 | 553 | |||
M33493.1 | - | 4 | 532 | |||
S55551.1 | - | 4 | 562 |
Gene Symbol: TPSAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003294.3 | NP_003285.2 | 5 | 607 | ||
GenBank mRNA | AF206665.1 | - | 4 | 565 | ||
AF206666.1 | - | 3 | 483 | |||
AF206667.1 | - | 3 | 369 | |||
BC038416.1 | - | 4 | 561 | |||
BC051852.1 | - | 4 | 560 | |||
BC065923.1 | - | 4 | 766 | 5752632 | ||
BC074974.2 | - | 5 | 588 | 30915512 | ||
BC074975.2 | - | 5 | 588 | 30915294 | ||
CR541836.1 | - | 4 | 555 | |||
FJ931118.1 | - | 3 | 404 | |||
M30038.1 | - | 4 | 572 | |||
M33491.1 | - | 4 | 551 | |||
M37488.1 | - | 4 | 560 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
