siRNA Details
Gene Symbol: TPSB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024164.5 | NP_077078.5 | 5 | 585 | ||
GenBank mRNA | AK308208.1 | - | 5 | 586 | ||
BC028059.1 | - | 4 | 574 | 5181815 | ||
BC029356.1 | - | 5 | 585 | 4695458 | ||
FJ931119.1 | - | 3 | 416 | |||
FJ931120.1 | - | 3 | 415 | |||
M33492.1 | - | 4 | 560 | |||
M33493.1 | - | 4 | 539 | |||
S55551.1 | - | 4 | 569 |
Gene Symbol: TPSAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003294.3 | NP_003285.2 | 5 | 614 | ||
GenBank mRNA | AF206665.1 | - | 4 | 572 | ||
AF206666.1 | - | 3 | 490 | |||
AF206667.1 | - | 3 | 376 | |||
BC038416.1 | - | 4 | 568 | |||
BC051852.1 | - | 4 | 567 | |||
BC065923.1 | - | 4 | 773 | 5752632 | ||
BC074974.2 | - | 5 | 595 | 30915512 | ||
BC074975.2 | - | 5 | 595 | 30915294 | ||
CR541836.1 | - | 4 | 562 | |||
FJ931118.1 | - | 3 | 411 | |||
M30038.1 | - | 4 | 579 | |||
M33491.1 | - | 4 | 558 | |||
M37488.1 | - | 4 | 567 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
