siRNA Details
Gene Symbol: TPSAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003294.3 | NP_003285.2 | 3 | 208 | ||
GenBank mRNA | BC038416.1 | - | 2 | 162 | ||
BC065923.1 | - | 2 | 367 | 5752632 | ||
BC074974.2 | - | 3 | 189 | 30915512 | ||
BC074975.2 | - | 3 | 189 | 30915294 | ||
CR541836.1 | - | 2 | 156 | |||
M33491.1 | - | 2 | 152 | |||
M37488.1 | - | 2 | 161 |
Gene Symbol: TPSB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC029356.1 | - | 3 | 179 | 4695458 | |
M33492.1 | - | 2 | 154 | |||
S55551.1 | - | 2 | 163 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
