siRNA Details
Gene Symbol: USP42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032172.2 | NP_115548.1 | 13 | 1684 | ||
XM_005249883.4 | XP_005249940.1 | 13 | 1631 | |||
XM_006715790.1 | XP_006715853.1 | 13 | 1631 | |||
XM_006715791.2 | XP_006715854.1 | 16 | 2137 | |||
XM_011515573.1 | XP_011513875.1 | 17 | 2196 | |||
XM_011515574.1 | XP_011513876.1 | 14 | 1720 | |||
XM_011515577.1 | XP_011513879.1 | 12 | 1349 | |||
XM_011515578.1 | XP_011513880.1 | 14 | 1771 | |||
GenBank mRNA | AJ601395.1 | - | 12 | 1561 | ||
AK022759.1 | - | 13 | 1628 | |||
AK301527.1 | - | 11 | 1156 | |||
AK309375.1 | - | 12 | 1425 | |||
AY618868.1 | - | 13 | 1628 | |||
BC060846.1 | - | 13 | 1623 | |||
BC132862.1 | - | 12 | 1581 | 40146884 | ||
BC136656.1 | - | 11 | 2514 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
