siRNA Details
Gene Symbol: KCNJ4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004981.1 | NP_004972.1 | 2 | 951 | ||
NM_152868.2 | NP_690607.1 | 2 | 1113 | |||
GenBank mRNA | BC032562.1 | - | 2 | 1123 | ||
BC039618.1 | - | 2 | 996 | |||
BC113506.1 | - | 1 | 905 | 8322558 | ||
BC113508.1 | - | 1 | 905 | 8322560 | ||
CR456507.1 | - | 2 | 921 | |||
S72503.1 | - | 2 | 952 | |||
U07364.1 | - | 2 | 951 | |||
U24056.1 | - | 2 | 1088 |

More Information
Other Species Targeted

Gene Ontology Categories:
