siRNA Details
Gene Symbol: LSM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014462.2 | NP_055277.1 | 2 | 495 | ||
NR_045492.1 | 2 | 495 | ||||
NR_045493.1 | 2 | 217 | ||||
GenBank mRNA | AF000177.1 | - | 2 | 249 | ||
AF147348.1 | - | 2 | 138 | |||
AJ238094.1 | - | 2 | 273 | |||
AK312159.1 | - | 2 | 248 | |||
BC001767.1 | - | 2 | 182 | 3353833 | ||
BP376421.1 | - | 2 | 458 | |||
BU521199.1 | - | 2 | 260 | |||
BU941962.1 | - | 2 | 220 | |||
KU178606.1 | - | 2 | 85 | |||
KU178607.1 | - | 2 | 85 |
