siRNA Details
Gene Symbol: BTN3A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242803.1 | NP_001229732.1 | 6 | 841 | ||
NM_006994.4 | NP_008925.1 | 11 | 1496 | |||
NM_197974.2 | NP_932078.2 | 10 | 1404 | |||
GenBank mRNA | AB209581.1 | - | 7 | 3024 | ||
AK223279.1 | - | 11 | 1431 | |||
AK223301.1 | - | 11 | 1415 | |||
AK291722.1 | - | 11 | 1480 | |||
AK301553.1 | - | 10 | 1388 | |||
BC015815.2 | - | 11 | 1433 | 3677284 | ||
BT007251.1 | - | 9 | 1253 | |||
KM244652.1 | - | 6 | 749 | |||
U90548.1 | - | 11 | 1424 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
