siRNA Details
Gene Symbol: HSD17B7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016371.3 | NP_057455.1 | 9 | 1421 | ||
GenBank mRNA | AF098786.2 | - | 9 | 1333 | ||
AF145023.1 | - | 9 | 1417 | |||
AJ249179.1 | - | 9 | 1346 | |||
AK022929.1 | - | 9 | 2440 | |||
AK290741.1 | - | 9 | 1390 | |||
AY358962.1 | - | 9 | 1329 | |||
BC065246.1 | - | 9 | 1392 | 6062803 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
