siRNA Details
Gene Symbol: DIMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014473.2 | NP_055288.1 | 1, 2 | 216 | ||
XM_011543323.2 | XP_011541625.1 | 1, 2 | 254 | |||
GenBank mRNA | AF102147.1 | - | 1, 2 | 165 | ||
AK292723.1 | - | 1, 2 | 244 | |||
AK299480.1 | - | 1, 2 | 206 | |||
BC002841.2 | - | 1, 2 | 213 | 3638494 | ||
BC010874.2 | - | 1, 2 | 177 | 3878818 | ||
BM749030.1 | - | 1, 2 | 226 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
