siRNA Details
Gene Symbol: PRSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002769.4 | NP_002760.1 | 5 | 714 | ||
XM_011516411.1 | XP_011514713.1 | 7 | 1410 | |||
GenBank mRNA | AK312199.1 | - | 5 | 708 | ||
BC128226.1 | - | 5 | 710 | 40114572 | ||
CR627200.1 | - | 1 | 3501 | |||
DQ778083.1 | - | 6 | 667 |
Gene Symbol: PRSS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303414.1 | NP_001290343.1 | 6 | 779 | ||
NM_002770.3 | NP_002761.1 | 5 | 737 | |||
NR_130149.1 | 5 | 663 | ||||
GenBank mRNA | BC030260.1 | - | 5 | 698 | ||
BC103997.2 | - | 5 | 703 | 40022560 | ||
BC103998.2 | - | 6 | 703 | 40022561 | ||
BC107784.1 | - | 5 | 714 | 6668396 | ||
DQ846872.1 | - | 7 | 694 | |||
DQ846873.1 | - | 6 | 739 | |||
M27602.1 | - | 5 | 703 |
Gene Symbol: PRSS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197097.2 | NP_001184026.2 | 6 | 1047 | ||
NM_001197098.1 | NP_001184027.1 | 5 | 689 | |||
NM_002771.3 | NP_002762.2 | 5 | 710 | |||
NM_007343.3 | NP_031369.2 | 5 | 919 | |||
XM_011517965.1 | XP_011516267.1 | 6 | 970 | |||
GenBank mRNA | AB298285.1 | - | 5 | 919 | ||
AB298286.1 | - | 5 | 689 | |||
AY052783.1 | - | 6 | 824 | |||
BC030238.1 | - | 5 | 709 | |||
BC069476.1 | - | 5 | 710 | 7262223 | ||
BC069494.1 | - | 5 | 710 | 7262235 | ||
D45417.1 | - | 5 | 697 | |||
X15505.1 | - | 5 | 710 | |||
X71345.1 | - | 5 | 737 | |||
X72781.1 | - | 5 | 740 |
Gene Symbol: PRSS3P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001296.3 | 6 | 780 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
